Learn More
Lenz Laborglasintrumente Receiver Adapters, Straight, With GL Thread
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
39.00 - 52.00
Produktcode | Merk | Standaard conische verbinding nr. | Prijs | Hoeveelheid en beschikbaarheid | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Merk | Standaard conische verbinding nr. | Prijs | Hoeveelheid en beschikbaarheid | |||||
10341190
|
Lenz Laborglasintrumente
05272014 |
Socket and Cone NS 29/32 |
43.00
1 stuk |
|
|||||
11516824
|
Lenz Laborglasintrumente
05272012 |
Socket and Cone NS 19/26 |
39.00
1 stuk |
|
|||||
11526824
|
Lenz Laborglasintrumente
05272013 |
Socket and Cone NS 24/29 |
52.00
Each |
|
|||||
Beschrijving
- Receiver adapters acc. to DIN 12594
- Straight
- With NS socket and NS cone
- With drain tip
- Vacuum connection: thread GL18
- Made of DURAN™ tubing
Specificatie
Duran Glass |