Learn More
Lenz Laborglasintrumente Receiver Adapters, Straight, With GL Thread
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Merk:  Lenz Laborglasintrumente 05272014
30.94 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 Bekijk meerdere types van dit product via de
Produktcode. 10341190
  • 43.00 / 1 stuk
Only null left
Toevoegen aan winkelmandje
Beschrijving
Beschrijving
  • Receiver adapters acc. to DIN 12594
  • Straight
  • With NS socket and NS cone
  • With drain tip
  • Vacuum connection: thread GL18
  • Made of DURAN™ tubing
Specificatie
Specificatie
Duran Glass
Socket and Cone NS 29/32