Learn More
Lenz Laborglasintrumente Receiver Adapters, Straight, With GL Thread
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Merk: Lenz Laborglasintrumente 05272013
32.81 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Beschrijving
- Receiver adapters acc. to DIN 12594
- Straight
- With NS socket and NS cone
- With drain tip
- Vacuum connection: thread GL18
- Made of DURAN™ tubing
Specificatie
Duran Glass | |
Socket and Cone NS 24/29 |