Learn More
Macherey-Nagel™ Blotting paper MN 218 B
Market leading paper quality for reliable biomolecule blottings – MN 218 B
39.70 - 268.00
Specificatie
Type | Blotting Paper |
---|---|
Aantal | 100 Pack |
Dikte (metrisch) | 0.36 mm |
Produktcode | Merk | Afmetingen (L x B) | Prijs | Hoeveelheid en beschikbaarheid | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Merk | Afmetingen (L x B) | Prijs | Hoeveelheid en beschikbaarheid | |||||
10307981
|
Macherey-Nagel™
742131 |
210 x 90 mm |
45.95
|
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10737811
|
Macherey-Nagel™
742113 |
570 x 460 mm |
230.00
|
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10539121
|
Macherey-Nagel™
742112 |
300 x 600 mm |
170.00
|
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10490813
|
Macherey-Nagel™
742111 |
580 x 600 mm |
268.00
|
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11922302
|
Macherey-Nagel™
742139 |
70 x 100 mm |
39.70
|
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Beschrijving
- Speed: Slow
Specificatie
Blotting Paper | |
100 Pack | |
0.36 mm |