Learn More
Waters Corporation Atlantis T3 Column
Atlantis T3 columns are universal, silica-based, reversed-phase C18 columns.
1155.00 - 1560.00
Specificatie
Max. druk | 6000 psi (415 Bar) |
---|---|
Partikelvorm | Spherical |
Poriegrootte | 100 Å |
Aantal | 1/Ea. |
USP-type | L1 |
Produktcode | Merk | Lengte (metrisch) | Diameter (metrisch) | Partikelgrootte | Prijs | Hoeveelheid en beschikbaarheid | |||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Merk | Lengte (metrisch) | Diameter (metrisch) | Partikelgrootte | Prijs | Hoeveelheid en beschikbaarheid | |||
13203140
![]() |
Waters Corporation
186003714 |
150 mm | 1 mm | 3 μm |
1455.00
|
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10442885
![]() |
Waters Corporation
186003717 |
50 mm | 2.1 mm | 3 μm |
1252.50
|
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10236983
![]() |
Waters Corporation
186003718 |
100 mm | 2.1 mm | 3 μm |
1357.50
|
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10382105
![]() |
Waters Corporation
186003722 |
100 mm | 3 mm | 3 μm |
1357.50
|
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10434835
![]() |
Waters Corporation
186003723 |
150 mm | 3 mm | 3 μm |
1455.00
|
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10601036
![]() |
Waters Corporation
186003726 |
50 mm | 4.6 mm | 3 μm |
1252.50
|
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10780775
![]() |
Waters Corporation
186003729 |
150 mm | 4.6 mm | 3 μm |
1455.00
|
Please sign in to purchase this item. Need a web account? Register with us today! | |||
13223150
![]() |
Waters Corporation
186003734 |
50 mm | 2.1 mm | 5 μm |
1207.50
|
Please sign in to purchase this item. Need a web account? Register with us today! | |||
13233150
![]() |
Waters Corporation
186003735 |
100 mm | 2.1 mm | 5 μm |
1297.50
|
Please sign in to purchase this item. Need a web account? Register with us today! | |||
13213160
![]() |
Waters Corporation
186003744 |
50 mm | 4.6 mm | 5 μm |
1207.50
|
Please sign in to purchase this item. Need a web account? Register with us today! | |||
13233160
![]() |
Waters Corporation
186003746 |
100 mm | 4.6 mm | 5 μm |
1297.50
|
Please sign in to purchase this item. Need a web account? Register with us today! | |||
13293130
![]() |
Waters Corporation
186003713 |
50 mm | 1 mm | 3 μm |
1252.50
|
Please sign in to purchase this item. Need a web account? Register with us today! | |||
13213140
![]() |
Waters Corporation
186003715 |
20 mm | 2.1 mm | 3 μm |
1155.00
|
Please sign in to purchase this item. Need a web account? Register with us today! | |||
13223140
![]() |
Waters Corporation
186003716 |
30 mm | 2.1 mm | 3 μm |
1207.50
|
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10110784
![]() |
Waters Corporation
186003719 |
150 mm | 2.1 mm | 3 μm |
1455.00
|
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10288783
![]() |
Waters Corporation
186003721 |
50 mm | 3 mm | 3 μm |
1252.50
|
Please sign in to purchase this item. Need a web account? Register with us today! | |||
13243140
![]() |
Waters Corporation
186003724 |
20 mm | 4.6 mm | 3 μm |
1155.00
|
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10332195
![]() |
Waters Corporation
186003725 |
30 mm | 4.6 mm | 3 μm |
1207.50
|
Please sign in to purchase this item. Need a web account? Register with us today! | |||
13263140
![]() |
Waters Corporation
186003727 |
75 mm | 4.6 mm | 3 μm |
1282.50
|
Please sign in to purchase this item. Need a web account? Register with us today! | |||
13273140
![]() |
Waters Corporation
186003728 |
100 mm | 4.6 mm | 3 μm |
1357.50
|
Please sign in to purchase this item. Need a web account? Register with us today! | |||
13213150
![]() |
Waters Corporation
186003733 |
30 mm | 2.1 mm | 5 μm |
1155.00
|
Please sign in to purchase this item. Need a web account? Register with us today! | |||
13243150
![]() |
Waters Corporation
186003736 |
150 mm | 2.1 mm | 5 μm |
1417.50
|
Please sign in to purchase this item. Need a web account? Register with us today! | |||
13263150
![]() |
Waters Corporation
186003738 |
50 mm | 3 mm | 5 μm |
1207.50
|
Please sign in to purchase this item. Need a web account? Register with us today! | |||
13273150
![]() |
Waters Corporation
186003739 |
100 mm | 3 mm | 5 μm |
1297.50
|
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10388074
![]() |
Waters Corporation
186003740 |
150 mm | 3 mm | 5 μm |
1417.50
|
Please sign in to purchase this item. Need a web account? Register with us today! | |||
13283150
![]() |
Waters Corporation
186003741 |
250 mm | 3 mm | 5 μm |
1560.00
|
Please sign in to purchase this item. Need a web account? Register with us today! | |||
13203160
![]() |
Waters Corporation
186003743 |
30 mm | 4.6 mm | 5 μm |
1155.00
|
Please sign in to purchase this item. Need a web account? Register with us today! | |||
13223160
![]() |
Waters Corporation
186003745 |
75 mm | 4.6 mm | 5 μm |
1245.00
|
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10196016
![]() |
Waters Corporation
186003747 |
150 mm | 4.6 mm | 5 μm |
1417.50
|
Please sign in to purchase this item. Need a web account? Register with us today! | |||
13243160
![]() |
Waters Corporation
186003748 |
250 mm | 4.6 mm | 5 μm |
1560.00
|
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17173232
![]() |
Waters Corporation
186005652 |
75 mm | 2.1 mm | 3 μm |
1282.50
|
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17183232
![]() |
Waters Corporation
186005653 |
75 mm | 3 mm | 3 μm |
1282.50
|
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Beschrijving
- Atlantis T3 columns are universal, silica-based, reversed-phase C18 columns that provide balanced retention of polar and hydrophobic molecules and should be your first choice when developing separations of polar and non-polar compounds.
Specificatie
6000 psi (415 Bar) | |
100 Å | |
L1 | |
Column |
Spherical | |
1/Ea. | |
Yes |