Learn More
Waters Corporation Atlantis T3 Column
Atlantis T3 columns are universal, silica-based, reversed-phase C18 columns.
Merk:  Waters Corporation 186003735
Encompass
 Bekijk meerdere types van dit product via de
Produktcode. 13233150
  • 1297.50 /
Beschrijving
Beschrijving
  • Atlantis T3 columns are universal, silica-based, reversed-phase C18 columns that provide balanced retention of polar and hydrophobic molecules and should be your first choice when developing separations of polar and non-polar compounds.
Specificatie
Specificatie
2.1 mm
6000 psi (415 Bar)
5 μm
1/Ea.
Yes
100 mm
Spherical
100 Å
L1
Column