Learn More
Waters Corporation Atlantis T3 Column
Atlantis T3 columns are universal, silica-based, reversed-phase C18 columns.
Merk:  Waters Corporation 186003725
Encompass
 Bekijk meerdere types van dit product via de
Produktcode. 10332195
  • 1207.50 /
Beschrijving
Beschrijving
  • Atlantis T3 columns are universal, silica-based, reversed-phase C18 columns that provide balanced retention of polar and hydrophobic molecules and should be your first choice when developing separations of polar and non-polar compounds.
Specificatie
Specificatie
4.6 mm
6000 psi (415 Bar)
3 μm
1/Ea.
Yes
30 mm
Spherical
100 Å
L1
Column