missing translation for 'onlineSavingsMsg'
Learn More

TRIM47 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

145.00€ - 410.00€

Specifications

Antigen TRIM47
Concentration 0.37 mg/mL
Applications Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16885215
View Documents
Proteintech
26885-1-AP-20UL
20 μL
145.00€
20µL
Please to purchase this item. Need a web account? Register with us today!
16875215
View Documents
Proteintech
26885-1-AP-150UL
150 μL
410.00€
150µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Synaptogyrins comprise a family of tyrosine-phosphorylated proteins with two neuronal (synaptogyrins 1 and 3) isoforms. Previous studies have indicated that synaptogyrins are involved in the regulation of neurotransmitter release. Synaptogyrin 1 is an integral membrane protein ubiquitously expressed in the nervous system. Synaptogyrin has four transmembrane domains and an extended cytoplasmic tail but it is not glycosylated. Synaptogyrin 1 is a tyrosine-phosphorylated by c-src and possibly other tyrosine kinases. The function of Synaptogyrin 1 is not well understood, but it may be a useful marker for nerve terminals and diagnosing neuroendocrine tumors.
Specifications

Specifications

TRIM47
Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Human
Q96LD4
91107
TRIM47 Fusion Protein Ag22868
Primary
-20°C
TRIM47
0.37 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
GOA, RING finger protein 100, RNF100, TRIM47, tripartite motif containing 47
TRIM47
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.