missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRIM47 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Merk: Proteintech 26885-1-AP-150UL
This item is not returnable.
View return policy
Beschrijving
Synaptogyrins comprise a family of tyrosine-phosphorylated proteins with two neuronal (synaptogyrins 1 and 3) isoforms. Previous studies have indicated that synaptogyrins are involved in the regulation of neurotransmitter release. Synaptogyrin 1 is an integral membrane protein ubiquitously expressed in the nervous system. Synaptogyrin has four transmembrane domains and an extended cytoplasmic tail but it is not glycosylated. Synaptogyrin 1 is a tyrosine-phosphorylated by c-src and possibly other tyrosine kinases. The function of Synaptogyrin 1 is not well understood, but it may be a useful marker for nerve terminals and diagnosing neuroendocrine tumors.Specificatie
TRIM47 | |
Polyclonal | |
Unconjugated | |
TRIM47 | |
GOA, RING finger protein 100, RNF100, TRIM47, tripartite motif containing 47 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
91107 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.37 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q96LD4 | |
TRIM47 | |
TRIM47 Fusion Protein Ag22868 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction