Learn More
Poulten & Graf GmbH Spare Piston for FORTUNA™ OPTIFIX™ BASIC Dispenser
Poulten & Graf GmbH Spare Piston for FORTUNA™ OPTIFIX™ BASIC Dispenser comes with a finger guard and volume setting system.
Merk: Poulten & Graf GmbH 101 00056
This item is not returnable.
View return policy
Beschrijving
- Solid PTFE encased
- With finger guard and volume setting system
Specificatie
300 mL | |
300 mL | |
1 | |
Spare parts for Dispensers | |
Reusable |
Replacement Piston for OPTIFIX BASIC Dispenser | |
OPTIFIX BASIC Dispenser | |
1 | |
Autoclavable | |
Non-sterile |