missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NCALD Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
145.00 - 410.00
Specificatie
Antigeen | NCALD |
---|---|
Concentratie | 0.51 mg/mL |
Toepassingen | Western Blot, Immunohistochemistry (Paraffin) |
Classificatie | Polyclonal |
Conjugaat | Unconjugated |
Produktcode | Merk | Aantal | Prijs | Hoeveelheid en beschikbaarheid | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Merk | Aantal | Prijs | Hoeveelheid en beschikbaarheid | |||||
16870884
|
Proteintech
12925-1-AP-20UL |
20 μL |
145.00
|
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16860884
|
Proteintech
12925-1-AP-150UL |
150 μL |
410.00
|
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Beschrijving
This gene encodes a member of the neuronal calcium sensor (NCS) family of calcium-binding proteins. The protein contains an N-terminal myristoylation signal and four EF-hand calcium binding loops. The protein is cytosolic at resting calcium levels; however, elevated intracellular calcium levels induce a conformational change that exposes the myristoyl group, resulting in protein association with membranes and partial co-localization with the perinuclear trans-golgi network. The protein is thought to be a regulator of G protein-coupled receptor signal transduction. Several alternatively spliced variants of this gene have been determined, all of which encode the same protein; additional variants may exist but their biological validity has not been determined.Specificatie
NCALD | |
Western Blot, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Rat, Mouse, Human | |
P61601, Q5PQN0, Q91X97 | |
52589, 553106, 83988 | |
NCALD Fusion Protein Ag3614 | |
Primary | |
-20°C | |
NCALD |
0.51 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
NCALD, neurocalcin delta | |
Ncald | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title