Learn More
NCALD Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Merk:  Proteintech 12925-1-AP-150UL
 Bekijk meerdere types van dit product via de
Produktcode. 16860884
  • 410.00 /
Explore more special offers
Beschrijving
Beschrijving
This gene encodes a member of the neuronal calcium sensor (NCS) family of calcium-binding proteins. The protein contains an N-terminal myristoylation signal and four EF-hand calcium binding loops. The protein is cytosolic at resting calcium levels; however, elevated intracellular calcium levels induce a conformational change that exposes the myristoyl group, resulting in protein association with membranes and partial co-localization with the perinuclear trans-golgi network. The protein is thought to be a regulator of G protein-coupled receptor signal transduction. Several alternatively spliced variants of this gene have been determined, all of which encode the same protein; additional variants may exist but their biological validity has not been determined.
Specificatie
Specificatie
NCALD
Polyclonal
Unconjugated
NCALD
NCALD, neurocalcin delta
Rabbit
Antigen Affinity Chromatography
RUO
52589, 553106, 83988
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.51 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P61601, Q5PQN0, Q91X97
Ncald
NCALD Fusion Protein Ag3614
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions
Product Suggestions
Video's
SDS
Documentatie
Documentatie
Certificates
Speciale aanbiedingen
Speciale aanbiedingen