Learn More
Ansell Microgard™ 2000 Standard SOCO/CSI Coveralls with Hood
Designed to allow water vapor (perspiration) to escape from the suit yet will withstand saturation of liquid chemicals. Filter 100% of particulates down to 0.01 microns in size.
Merk: Ansell WH20B-00128-04
330.18 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Beschrijving
- Self adhesive pockets can be positioned anywhere on the garment to secure equipment
- Finger loops prevent sleeve movement when working above your head
- Protection from biological agents in the highest performance class according to EN14126
- Anti-static according to EN 1149-5
- Elasticated hood, wrists, waist and ankles
- 2-way front zipper with resealable storm flap
- 2-piece hood with chinstrap
- Dual finger loops
- Includes 2 pockets
Specificatie
General Purpose Coveralls | |
Unisex | |
White | |
Elastic | |
Elastic | |
2-panel with Chin Strap | |
Bound | |
Cat III Type 5-B, 6-B, EN1073-2, EN14126, EN1149-1, DIN 32781 | |
WH20-B-00-128 | |
Includes Hood with Chinstrap, Dual Fingerloops |
Large | |
Microporous Polyethylene Film | |
Single-use | |
Elastic | |
Hooded | |
Front Zipper with Storm Flap | |
Non-sterile | |
Hood with chinstrap, dual finger loops | |
2 |