Learn More
Nickel Electro™ Hinged Gable Lid
View samples better with a semi-transparent lid. Nickel Electro™ Hinged Gable Lid maximizes bath area while using its shape to drip condensation back into the bath.
Merk:  Nickel Electro™ GL1-14
113.26 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 Bekijk meerdere types van dit product via de
Produktcode. 12768888
  • 116.00 / 1 stuk
Only null left
Toevoegen aan winkelmandje
Inclusief: dimForUseWithEquipment
Vereist : ForUseWithApplication
Beschrijving
Beschrijving
  • Lightweight
  • Semi-transparent
  • Drips condensation back into bath
Transparent lid, hinged, high profile, handle
Specificatie
Specificatie
14 L Unstirred Baths
Reducing Water Evaporation, Keeping Samples and Water Free From Contamination and Saves Energy