Learn More
Sampling Systems™ Graduated SteriWare Beakers
This range of graduated beakers are crucial in the production of drugs, chemicals and cosmetics. Individually bagged to prevent cross contamination, these beakers also have a pouring spout to ensure the convenient handling of liquids.
61.45 - 154.00
Specificatie
Materiaal | Polypropylene |
---|---|
Autoclaveerbaar | Autoclavable |
Voor eenmalig gebruik | Single-use |
Graduated | Yes |
Certificeringen/conformiteit | FDA |
Produktcode | Merk | Inhoud (metrisch) | Steriliteit | Prijs | Hoeveelheid en beschikbaarheid | ||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Merk | Inhoud (metrisch) | Steriliteit | Prijs | Hoeveelheid en beschikbaarheid | ||||
17962052
|
Sampling Systems™
8008P-25 |
25 mL | Non-sterile |
61.45
Pack of 50 |
|
||||
17982052
|
Sampling Systems™
8008P-25S |
25 mL | Sterile |
119.00
Pack of 50 |
|
||||
17922062
|
Sampling Systems™
8008P-50 |
50 mL | Non-sterile |
62.95
Pack of 50 |
|
||||
17912062
|
Sampling Systems™
8008P-50S |
50 mL | Sterile |
122.00
Pack of 50 |
|
||||
17932052
|
Sampling Systems™
8008P-100 |
100 mL | Non-sterile |
65.50
Pack of 50 |
|
||||
17972052
|
Sampling Systems™
8008P-100S |
100 mL | Sterile |
130.00
Pack of 50 |
|
||||
17902062
|
Sampling Systems™
8008P-250 |
250 mL | Non-sterile |
99.00
Pack of 40 |
![]() |
||||
17992052
|
Sampling Systems™
8008P-250S |
250 mL | Sterile |
154.00
Pack of 40 |
|
||||
17992112
|
Sampling Systems™
8008P-2000 |
2 L | Non-sterile |
61.55
Pack of 15 |
![]() |
||||
17869965
|
Sampling Systems™
8008P-2000S |
2 L | Sterile |
122.00
Pack of 15 |
|
||||
Beschrijving
- Made from food grade, FDA and EC compliant polypropylene
- Available sterile and non-sterile
- Wide range of sizes
- 100% manufactured and packed in a cleanroom
Specificatie
Polypropylene | |
Single-use | |
FDA | |
Low Form |
Autoclavable | |
Yes | |
Molded Scale |