Learn More
Sampling Systems™ Graduated SteriWare Beakers
This range of graduated beakers are crucial in the production of drugs, chemicals and cosmetics. Individually bagged to prevent cross contamination, these beakers also have a pouring spout to ensure the convenient handling of liquids.
Merk: Sampling Systems™ 8008P-50S
111.92 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Beschrijving
- Made from food grade, FDA and EC compliant polypropylene
- Available sterile and non-sterile
- Wide range of sizes
- 100% manufactured and packed in a cleanroom
Specificatie
50 mL | |
Sterile | |
Single-use | |
FDA | |
Low Form |
Polypropylene | |
Autoclavable | |
Yes | |
Molded Scale |