Learn More
Kimble Chase™ Glass Tube for Potter-Elvehjem™ Tissue Grinder
Replacement Tube for Kimble™ Potter-Elvehjem™ Tissue Grinder.
Merk: Kimble Chase™ 885752-0022
52.31 EUR valid until 2025-06-13
Use promo code "24123" to get your promotional price.
This item is not returnable.
View return policy
Beschrijving
Specificatie
8 mL | |
Tube | |
22 |
Glass, PTFE | |
For Potter-Elvehjem Tissue Grinder | |
PTFE |