Learn More
Ansell™ AlphaTec™ Solvex™37-186
High-comfort, chemical resistant glove for a wide range of applications
Merk: Ansell™ 37186080
This item is not returnable.
View return policy
Beschrijving
- Available in a wide selection of lengths, thicknesses, sizes and linings, the Ansell Sol-Vex™ nitrile glove is designed to deliver optimal results in wet or dry work environments where chemical resistance is crucial. Fully reusable, with an unequaled abrasion resistance, it provides superb comfort for the wearer.
- Sol-Vex™ nitrile gloves are the ideal choice for safe handling in a wide range of work environments where harsh chemicals are present
- The unlined version of the glove has no inner cotton flocking, reducing risk of lint contamination and making it ideal for production areas sensitive to the introduction of external impurities.
- The reversed lozenge finish further enhances levels of grip.
- The sandpatch finish gives the glove a smooth surface that decreases indirect costs thanks to fewer rejects of fragile parts.
Specificatie
Nitrile | |
Category III | |
Index | |
Nitrile | |
1 | |
Sandpatch | |
Specific | |
Powder Free | |
8 | |
37-186 | |
Chemical and Liquid Protetion | |
Straight | |
Polybag/cardboard box | |
1 Pair/bag, 12 Pair/carton | |
Solvex™ | |
460 mm | |
82.5 g |
EN ISO 374-1:2016/Type A/AGJKLPT, EN ISO 374-5:2016 Bacteria, EN388:2016/4102X | |
Nitrile | |
High-comfort, chemical resistant glove for a wide range of applications | |
Green | |
Reusable | |
Industrial grade | |
Yes | |
Immersion Gloves | |
Non-sterile | |
0.65 mm | |
Green | |
37-186 | |
2 | |
Chemical and Liquid Protection | |
460 x 104 mm | |
104 mm |