Learn More
Buerkle™ Polypropylene Container Connector
Container connectors with outer thread for the easy mounting of thread in the sides of containers.
Merk:  Buerkle™ 8533-0034
 Bekijk meerdere types van dit product via de
Produktcode. 10147653
  • 16.85 / 1 stuk
Explore more special offers
Toevoegen aan winkelmandje
Beschrijving
Beschrijving
  • Polypropylene construction
  • Complete with EPDM seals
Specificatie
Specificatie
EU Foodstuffs
Autoclavable
Ball Valve
18 mm
Container Connection
Outer thread
Polypropylene
3/4 in.
White
Product Suggestions
Product Suggestions
Video's
SDS
Documentatie
Documentatie
Certificates
Speciale aanbiedingen
Speciale aanbiedingen