Learn More
Buerkle™ Polypropylene Container Connector
Container connectors with outer thread for the easy mounting of thread in the sides of containers.
Merk: Buerkle™ 8533-0034
This item is not returnable.
View return policy
Beschrijving
- Polypropylene construction
- Complete with EPDM seals
Specificatie
EU Foodstuffs | |
Autoclavable | |
Ball Valve | |
18 mm | |
Container Connection |
Outer thread | |
Polypropylene | |
3/4 in. | |
White |