Learn More
MARK1 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Merk: Proteintech 21552-1-AP-20UL
This item is not returnable.
View return policy
Beschrijving
The MARK1 antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human MARK1. This antibody recognizes human, mouse, rat antigen. The MARK1 antibody has been validated for the following applications: IHC, IP, WB, ELISA analysis.
Specificatie
MARK1 | |
Polyclonal | |
Unconjugated | |
20 μL | |
4139 | |
IgG |
Western Blot, Immunoprecipitation, Immunohistochemistry, ELISA | |
360 μg/mL | |
Rabbit | |
Primary | |
Human, Rat, Mouse |