Learn More
Karl Hecht™ Erlenmeyer Flask with Hollow NS Glass Stopper
Conical, flat-bottomed flask well suited to mixing by swirling. Karl Hecht™ Erlenmeyer Flask with Hollow NS Glass Stopper is constructed of durable Duran™ borosilicate 3.3 glass.
Merk: Karl Hecht™ 41918012
This item is not returnable.
View return policy
Beschrijving
- Durable Duran™ borosilicate 3.3 glass.
- Graduated.
- Reinforced ground neck marked with standard ground size.
- Hollow NS glass stopper.
Specificatie
Glass | |
Borosilicate Glass (3.3) | |
Yes | |
NS 29/32 |
50 mL | |
NS 29/32 | |
NS Stopper |