Learn More
DHRS3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Merk: Proteintech 15393-1-AP-20UL
This item is not returnable.
View return policy
Beschrijving
DHRS3 catalyze the oxidation/reduction of a wide range of substrates, including retinoids and steroids.Specificatie
DHRS3 | |
Polyclonal | |
Unconjugated | |
DHRS3 | |
DD83.1, DHRS3, RDH17, retSDR1, Rsdr1, SDR1, SDR16C1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
20148, 313689, 9249 | |
-20°C | |
Liquid |
Western Blot, Immunoprecipitation | |
0.25 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O75911, O88876 | |
Dhrs3 | |
DHRS3 Fusion Protein Ag7589 | |
20 μL | |
Primary | |
Rat, Mouse, Human | |
Antibody | |
IgG |