Learn More
ZG16 Mouse anti-Human, Unconjugated, Clone: 1A7B9, Proteintech
Mouse Monoclonal Antibody
Merk: Proteintech 67389-1-IG-20UL
This item is not returnable.
View return policy
Beschrijving
The ZG16 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human ZG16. This antibody recognizes human, mouse, pig, rat antigen. The ZG16 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specificatie
ZG16 | |
Monoclonal | |
Unconjugated | |
20 μL | |
653808 | |
IgG1 |
Western Blot, Immunohistochemistry, ELISA | |
1A7B9 | |
Mouse | |
Primary | |
Human, Mouse, Rat, Pig |