Learn More
ZG16 Mouse anti-Human, Unconjugated, Clone: 1A7B9, Proteintech
Mouse Monoclonal Antibody
Merk:  Proteintech 67389-1-IG-150UL
 Bekijk meerdere types van dit product via de
Produktcode. 16869985
  • 410.00 /
Beschrijving
Beschrijving
The ZG16 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human ZG16. This antibody recognizes human, mouse, pig, rat antigen. The ZG16 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specificatie
Specificatie
ZG16
Monoclonal
Unconjugated
150 μL
653808
IgG1
Western Blot, Immunohistochemistry, ELISA
1A7B9
Mouse
Primary
Human, Mouse, Rat, Pig