Learn More
YWHAB Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Merk: Proteintech 10936-1-AP-20UL
This item is not returnable.
View return policy
Beschrijving
This antibody was raised against the N-terminal region of human YWHAB.
14-3-3, a family of acidic and soluble proteins, highly conserved in amino acid sequences from yeast to mammals, is expressed in alleukaryotic cells. Seven isoforms encoded by seven distinct genes are identified in mammals and forms homo-and hetero-dimericcup-shaped structures. As 14-3-3 is interacted with more than 100 binding partners, it regulates key proteins involved in various biological processes such as signal trans-duction, cell cycle, transcriptional control, cell proliferation, apoptosis, and ion channel physiology. Most 14-3-3 requires phosphorylation of serine or threonine residues in the target sequence. This protein is abundantly expressed in the brain and has been detected in the cerebrospinal fluid of patients with different neurological disorders.Specificatie
YWHAB | |
Polyclonal | |
Unconjugated | |
YWHAB | |
14 3 3, 14 3 3 alpha, 14 3 3 protein beta/alpha, YWHAB | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
54401, 56011, 7529 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.4 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P31946, P35213, Q9CQV8 | |
Ywhab | |
YWHAB Fusion Protein Ag1383 | |
20 μL | |
Primary | |
Rat, Mouse, Human | |
Antibody | |
IgG |