missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Schneider Gerd™ Flexibele apothekersspatel met houten handgreep
Use this spatula for a variety of pharmaceutical applications. Schneider Gerd™ Wooden Handled Flexible Pharmacist Spatulas are ideal for use with a wide range of compounds.
Merk: Schneider Gerd™ 8613
This item is not returnable.
View return policy
Beschrijving
- Spatulas are ideal for working with powders, stirring and mixing
- Perfect for miniscule amounts of chemicals
- Polished stainless steel implements are effortlessly cleaned and corrosion resistant
Specificatie
Apothekersspatel | |
Stainless Steel | |
Houten handgreep | |
150 mm |
22 mm | |
Hout | |
Hout |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction