Learn More
Poulten & Graf GmbH VOLAC FORTUNA™ Bulb Pipette with 2 Marks and Blue Graduations
Poulten & Graf GmbH VOLAC FORTUNA™ Bulb Pipette with 2 Marks and Blue Graduations allow accurate measurements of a volume of solution.
Brand:  Poulten & Graf GmbH 1 2322902
 View more versions of this product
Product Code. 10639942
Description
Description
  • With 2 marks
  • Blue graduations
  • Conformity approved
  • With batch certificate
Specifications
Specifications
EN ISO 648
Soda Lime Glass
Yes
Ring marks
0.01 mL
Two-mark Bulb Pipette
AS
Non-sterile
Blue
5
3 mL
Reusable