Learn More
Poulten & Graf GmbH VOLAC FORTUNA™ Bulb Pipette with 2 Marks and Blue Graduations
Poulten & Graf GmbH VOLAC FORTUNA™ Bulb Pipette with 2 Marks and Blue Graduations allow accurate measurements of a volume of solution.
Merk: Poulten & Graf GmbH 1 2324502
This item is not returnable.
View return policy
Beschrijving
- With 2 marks
- Blue graduations
- Conformity approved
- With batch certificate
Specificatie
EN ISO 648 | |
Soda Lime Glass | |
Yes | |
Ring marks | |
0.05 mL | |
Two-mark Bulb Pipette |
AS | |
Non-sterile | |
Blue | |
5 | |
50 mL | |
Reusable |