Learn More
Contec™ Anticon™ Gold Standard Weight Cleanroom Wipes
Clean sensitive optics in ISO Class 1 to 4 applications. Contec™ Anticon™ Gold Standard Weight Cleanroom Wipes can be used in many applications from spill control to critical chamber cleaning. 100% continuous filament polyester material is chemically modified for superior cleanliness and sorbency.
Merk: Contec™ 495353-903
415.96 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Beschrijving
- Ideal for wiping down sensitive optics, excellent for superior spill control, for ISO Class 1 to 4 applications
- Engineered to deliver maximum performance at a superior value
- These all-purpose wipes can be used in many applications from spill control to critical chamber cleaning
- Double-knitted from 100% continuous filament polyester in a no-run interlock pattern
- Polyester yarn is chemically engineered for maximum sorbency and cleanliness
- Unique sealing technology truly seals wipe edges, allowing the use of 100% of the wipe surface
- Wipes feature low sub-micron particle and fiber counts, low extractable levels, and fast absorption rate for quick pickup of spills 100% continuous filament polyester chemically modified for superior cleanliness and sorbency
- Excellent wipe for critical cleaning, wiping, and spill control
Compliance
Tested in accordance with IEST Recommended Practices for the Evaluation of Wiping Materials for Critical Environments.
Specificatie
Cleanroom Wipe | |
23 cm | |
23 cm | |
ISO Class 1 to 4 | |
Ideal for wiping down sensitive optics, excellent for superior spill control, for ISO Class 1 to 4 applications |
9 in. | |
9 in. | |
Polyester | |
22.86 x 22.86 cm (9 x 9 in.) | |
2 Bags x 75 Wipes (Bulk) |