Learn More
Metro™ Super Erecta™ SiteSelect™ Posts
For use with Metro Super Erecta, Super Adjustable Super Erecta and Super Erecta Pro wire shelving systems
Merk:  Metro™ 63UPS
 Bekijk meerdere types van dit product via de
Produktcode. 15966741
  • 115.00 / 1 stuk
Only null left
Toevoegen aan winkelmandje
Beschrijving
Beschrijving
  • Stationary posts are fitted with adjustable leveling bolts to compensate for uneven surfaces.
  • Posts are available in chrome, type 304 stainless steel, and metroseal 3.
  • Metroseal 3 includes Microban™ antimicrobial product protection and has a 12 year warranty against rust and corrosion.
  • Mobile posts come without leveling bolt assembly for use with stem casters.
  • Grooved at 1 in. (2.5 cm) intervals.
  • Posts are double-grooved every 8 in. (20.3 cm) for easy identification.
Informatie over bestelling
Height includes leveling bolt and cap. Shelves and posts sold separately.
Specificatie
Specificatie
Mobile Shelving Post
Metro Super Erecta Wire Shelving System
157 cm
Stainless Steel
61.81 in.
Mobile