Learn More
Fisherbrand™ Universal Absorbent Roll
Fisherbrand™ Universal Absorbent Roll absorbs industrial liquids including oils, water, solvents and coolants.
Merk:  Fisherbrand™ FBEA555E
223.60 EUR valid until 2025-06-30
Use promo code "24156" to get your promotional price.
Produktcode. 15843720
  • 317.00 /
Only null left
Toevoegen aan winkelmandje
Beschrijving
Beschrijving
  • One sided spunbound offers durability and low lint on one side
  • Multiple perforations for ease of use and to minimize waste
  • Multi-use maintenance sorbent that can be used in five different ways
    • Soc to catch leaks and drips around machines
    • Wipe to clean machines or machine parts
    • Pad to soak up small spills
    • Roll to keep the walkways safe
    • Pillow under persistent drip sources
Specificatie
Specificatie
Grijs
2-laags, spinvlies/smeltspin polypropyleen, gebonden
Machinenevel, lekkages, spatten, in opslagruimten voor vaten en vloeistoffen, werkstations, rond en onder lekkende machines.
2-ply
150ft.
14,96in.
2,99in
15,87 lb.
92L
Universele adsorptiemiddelen
1 rol
46m x 38cm
46m
38cm
76mm
7,2 kg