Learn More
Eppendorf Reference™ 2-pipetten met variabel volume
Minimaliseer het risico van aerosolvervuiling en verminder duimklachten met deze pipetten met variabel volume. De Eppendorf™ Reference™ 2 Pipetten met Variabel Volume zijn ideaal voor het pipetteren en mengen van vloeistoffen, voor het vullen van platen, voor gels en reageerbuisjes, voor fase-extractie en voor het verwijderen van bovendrijvend materiaal.
Merk: Eppendorf 4924000096
375.25 EUR valid until 2025-12-31
Use promo code "24314" to get your promotional price.
Beschrijving
- Integrated pipetting system with internal RFID technology
- Serial number is printed on multiple components of pipette
- Color coded and volume labeled pipette for quick identification of volume size/tip size
- Round upper part of pipette makes it possible to work in every position
- Four-digit display for exact, quick and secure volume setting (volume lock included)
- Spring-loaded tip cone reduces effort during operation/ejection
- Sturdy upper handle, movable parts and external edges made from stainless-steel make pipette robust
- High flexibility with ability to remove single cones
- Ensures continuous identical pipet alignment during work process
- Fully autoclavable pipet with smooth surface and few rough edges
- Single-button operation
- Has Spring-loaded tip cone, movable parts, stainless steel external edges with customizable labeling area
- Ergonomic handling with reduced operating effort
- Simple and quick tip ejection with active aerosol reduction
- Reduce repetitive strain injuries by not having to move your thumb sideways
- Haptic feedback before tip ejection
- High precision and accuracy provides reliable pipetting results
- User friendly secondary adjustment
Specificatie
Ja | |
Pipette Tip 2500 μL | |
Reference 2 | |
0.25-2.5 mL | |
epT.I.P.S Sample Bag with 10 Pipette Tips | |
Red |
Rood | |
1 | |
Pipette | |
Voor gebruik met pipetpunten van 2500 μL | |
2 μl |