Learn More
Contec™ CONTEXT™ Cleanroom Paper and Notebooks
CONTEXT™ Cleanroom Documentation products have been developed to provide cleanrooms with paper and notebooks that reduce the risk of contamination. Available in 8.5 x 11 in., A4, and 11 x 17 in. sizes.
Merk: Contec™ CNTXA4B
182.77 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Beschrijving
CONTEXT™ paper uses Munising Bond material made from synthetic polymer-saturated cellulose that provides optimal smoothness and natural enhanced writeability.- High brightness, high opacity
- Ultra low particle generation
- Ultra low metallic ion content
- High tear and tensile strength
- Heat resistant
- Compatible with standard and high-speed laser printers
- Compatible with wet and dry copy systems
- Ideal for recording notes and instructions
Specificatie
Munising Bond Material from Synthetic Polyer-saturated Cellulose | |
8.3 in. | |
Blue | |
210 x 297 mm | |
Single-use | |
Standard and high-speed laser printers; wet and dry copy systems | |
Cleanroom Paper | |
A4 | |
22 lb. |
No | |
11.7 in. | |
Blue | |
Autoclavable | |
Recording notes and work instructions | |
Yes | |
250 Sheets/Pk., 10 Packs/Cs. | |
Non-sterile | |
83 gsm |