Learn More
Quickfit™ kolf met platte bodem en korte hals, borosilicaatglas
Manufactured from borosilicate glass. Quickfit Jointed Ware™ Quickfit™ Borosilicate Glass Short Neck Flat Bottom Flask complies with ISO 1773 and ISO 4797.
Merk: Quickfit FF1L/5S
This item is not returnable.
View return policy
Beschrijving
Manufactured from borosilicate glass. Quickfit Jointed Ware™ Quickfit™ Borosilicate Glass Short Neck Flat Bottom Flask complies with ISO 1773 and ISO 4797.
- Complies with ISO 1773 and ISO 4797
- With highest quality Quickfit™ ground glass socket
- Borosilicate glass for highest resistance to thermal shock and chemical corrosion
- Flat base allows use with hotplates
Specificatie
ISO 1773, ISO 4797 | |
Borosilicate Glass | |
Short | |
Flat |
1000 mL | |
175 mm | |
34/35 |