Learn More
Invitrogen™ PCR Master Mix Starter Pack
The PCR Master Mix Starter Pack is composed of three different PCR master mixes for easy evaluation of their suitability for use in your research.
Merk:  Invitrogen™ A31021
285.35 EUR valid until 2025-06-30
Use promo code "24108" to get your promotional price.
Produktcode. 15545053
  • 451.00 /
Explore more special offers
Beschrijving
Beschrijving
The PCR Master Mix Starter Pack is composed of three different PCR master mixes for easy evaluation of their suitability for use in your research. These master mixes provide our best DNA polymerases for routine PCR, hot-start PCR, and high-fidelity PCR needs. Thermo Scientific DreamTaq Green PCR Master Mix contains an enhanced Taq DNA polymerase optimized for all standard PCR applications. Invitrogen Platinum II Hot Start PCR Master Mix contains hot-start Taq DNA polymerase for specific, robust, and reliable PCR. Invitrogen SuperFi II PCR Master Mix features superior performance and an error rate 300 times lower than that of Taq. It is an excellent choice for cloning, mutagenesis, and other applications requiring high fidelity.
Master mixes included in this starter pack:
  • 2X DreamTaq Green PCR Master Mix (2X), 200 reactions (Cat. No. K1081)
  • 2X Platinum II Hot Start PCR Master Mix (2X), 50 reactions (14000012)
  • Platinum SuperFi II PCR Master Mix, 100 reactions (12368010)
TRUSTED_SUSTAINABILITY
Specificatie
Specificatie
• 2X DreamTaq Green PCR Master Mix, 200 reactions
• 2X Platinum II Hot-Start PCR Master Mix, 50 reactions
• 2X Platinum SuperFi II PCR Master Mix, 100 reactions
Store all components at -20°C.
PCR Master Mix Starter Pack
1 pack
Hot-start PCR
Kit
Platinum II Taq Hot Start DNA Polymerase
PCR Master Mix Starter Pack
Dry Ice
350 Reactions
DNA