Learn More
IKA™ Eurostar Digital Laboratory Stirrer
Automatically adjusts the stirring speed through microprocessor controlled technology within the speed range of 0/30 - 2000 rpm. IKA™ Eurostar Digital Laboratory Stirrer includes safety circuits to ensure automatic cut-off in anti-stall or overload conditions. Variations are automatically adjusted to guarantee a constant speed even with changes in viscosities of the sample.
Merk: IKA™ 0004442000
930.30 EUR valid until 2025-04-30
Use promo code "24119" to get your promotional price.
Beschrijving
Laboratory stirrer designed for simple stirring tasks.- Automatically adjusts the speed through microprocessor controlled technology within the speed range of 0/30 - 2000 rpm.
- Safety circuits installed ensures automatic cut-off in anti-stall or overload conditions.
- Continuous comparison of shaft speed to desired speed is maintained and variations are adjusted automatically to guarantee a constant speed even with changes in viscosities of the sample.
- Digital speed display
- Infinitely adjustable speed
- Push-through agitator shafts
- Overload protection
- Short-term overload operation
- Slim casing
- Quiet operation
- Error code display
Specificatie
0/30 to 2000 rpm | |
Digital | |
208 mm | |
248 mm | |
86 mm | |
DIN EN 60529 | |
Eurostar 20 digital | |
20 Ncm | |
10,000 mPas |
15 L | |
100/115/230 V | |
11 mm | |
16 mm | |
50/60 Hz | |
Alu-Cast Coating/Thermoplastic Polymer | |
IP40 | |
Stirrer | |
4.4 kg |