Learn More
Cytiva illustra™ Ready-To-Go™ GenomiPhi V3 DNA-amplificatiekit
Zeer efficiënte en representatieve vermeerdering van hele genoom met een opbrengst van 40 tot 60 μg van nanogramhoeveelheden van een DNA-monster. GE Healthcare Lifescience™ illustra™ Ready-To-Go™ GenomiPhi V3 DNA-amplificatiekit maakt deel uit van de Phi29 DNA-polymerasegroep van producten.
Merk: Cytiva 25-6601-97
3090.35 EUR valid until 2025-12-31
Use promo code "24339" to get your promotional price.
Beschrijving
p>- Representative amplification of the whole genome.
- Preformulated, predispensed, single-dose, lyophilized cakes for rapid reaction set-up and greater reproducibility.
- Quick and simple automation-friendly protocol; no thermal cycler required.
- Shipping and storage at room temperature provides easy access and reduced carbon footprint.
- Yields high-quality DNA (e.g. for high-throughput genotyping and DNA archival).
- The reaction mixture is provided as predispensed, single-dose, lyophilized cakes in either strips of 8 tubes, or 96-well plates.
- Amplification is highly uniform over the entire genome so that locus representation remains extremely close to the original DNA sample.
- Amplification is carried out with very high fidelity due to Phi29 DNA polymerase proofreading activity.
- The average product length is over 10kb.
- Procedure is quick, simple and does not require a thermocycler.
- A typical DNA yield of 12 to 20μg DNA can be achieved in less than two hours from only 10ng of genomic DNA input.
- The starting material for GenomiPhi reactions can be purified DNA from any commercial kit or homebrew method, or a nonpurified cell lysate may be used.
- Protocols for the amplification of DNA from blood lysates and dried blood spots are included in the manual.
- GenomiPhi amplified DNA is suitable for various applications such as genotyping (SNP, STR, array CGH), cloning, sequencing, and DNA archival.
Specificatie
Store the kit at ambient temperature.Once opened, completely reseal the pouch containingthe Ready-To-Go GenomiPhi V3 DNA amplification kit cakes. To ensure maximum shelf life and optimal performance, storage in a desiccator is recommended.Store recons | |
Genomic DNA Amplification | |
illustra Ready-To-Go GenomiPhi V3 DNA Amplification Kit 5x96 Reactions | |
1.5-2 h |
illustra Ready-To-Go GenomiPhi V3 DNA Amplification Kit offers highly efficient and representative whole-genome amplification with 12 to 20 μg yield from nanogram amounts of DNA sample. | |
480 | |
> 10 kb |