Learn More
Berkshire™ Cleanroom-doekje van polyester en cellulose
BIOPHARMA
Use these wipes for cleaning and maintenance in laboratories as well as aerospace and defense, microelectronics, food processing, pharmaceutical and biomedical device manufacturing environments. Berkshire™ Durx™ 670 General Purpose Wipes feature exceptional strength and absorbency.
Merk:  Berkshire™ DR670.0909.20
Produktcode. 12877143
  • 36.00 /
Only null left
Edge
Toevoegen aan winkelmandje
Beschrijving
Beschrijving
  • Manufactured without the use of binders or surfactants, ion levels and organic extractables remain low
  • The same wipe material can be provided in pre-wetted formats for reduced VOC emissions, increased convenience, increased productivity, improved solvent control and cleaning protocol repeatability and reduced costs
  • For aseptic processing areas, the same wipe material can be provided in a gamma irradiated format, validated sterile to a 10–6 sterility assurance level
  • Dry and pre-wetted versions available
  • FDA 21 CFR – Food contact compliant
Specificatie
Specificatie
Cleanroom-doekje
230 mm
Messnede
230 mm
Polyester en cellulose
Pakket 300