Learn More
Berkshire International™ Durx™ 770 Cleanroom Wipes, Class 100, ISO Class 5
BIOPHARMA
Ultra-low particle cleanroom wipes ideally suited for critical and small parts cleaning. Berkshire International™ Durx™ 770 Cleanroom Wipes, Class 100, ISO Class 5 are highly absorbent and exhibit high wet strength.
Merk:  Berkshire International™ DR770.0909.20
Produktcode. 12867143
  • 50.65 /
Only null left
Toevoegen aan winkelmandje
Beschrijving
Beschrijving
  • Suitable for cleaning and wiping.
  • Particularly suited for heavy-duty use, regular maintenance tasks, photo resist applications, and around etching baths.
  • Wipe can be used for cleaning and maintenance in laboratories, aerospace, electronics, food processing, photo processing, and semiconductor, circuit board, pharmaceutical, and biomedical device manufacture.
  • Non-woven cellulose and polyester fibers, specially processed to reduce particle generation and increase absorbancy while retaining high wet strength.
Specificatie
Specificatie
Cleanroom Wiper
High
Class 100, ISO Class 5
23 x 23 cm
300 Pk.
Polyester Cellulose
Autoclavable
Non-woven
Cleaning and maintenance in laboratories, aerospace, electronics, food processing, photo processing, and semiconductor, circuit board, pharmaceutical, and biomedical device manufacture