Learn More
R&L Enterprises™ Bunsenbrander, aardgas
An excellent solution for heating, sterilization and combustion in the laboratory. R&L Enterprises™ 13mm Natural Gas Bunsen Burners have a typical gas consumption of 107.5L per hour.
Merk:  R&L Enterprises™ 4006/2
Produktcode. 12832154
  • 151.84 / 5 stuks
Only null left
Toevoegen aan winkelmandje
Beschrijving
Beschrijving
  • For use with natural (methane) gas
  • Temperature is achieved at the tip of the blue flame cone
  • With air regulator and rifled gas inlet tube
  • Constructed using a heavy pressure diecast base painted in chemically resistant blue acrylic finish
  • Tested for flame quality and leakage prior to packing
Specificatie
Specificatie
Bunsenbrander
13 mm
145 mm
1 kg
Natuurlijk/methanol
9 mm
5 per pak