Learn More
DWK Life Sciences Kontes™ rechthoekige TLC ontwikkeltanks met deksel
Molded glass tanks for development of any size TLC plate up to 20 x 20cm
Merk:  DWK Life Sciences 4161850000
 Bekijk meerdere types van dit product via de
Produktcode. 12766728
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Bekijk hier alternatieve producten »
Beschrijving
Beschrijving
Molded glass tanks for development of any size TLC plate up to 20 x 20cm
  • Tanks: K416180-0000, K416185-0000, K416190-0000
  • Lids: K416181-0000, K416186-0000
TRUSTED_SUSTAINABILITY
Specificatie
Specificatie
Nonslip
Developing Tank
4 x 11.75 x 10.75 in. (10.16 x 29.8 x 27 cm)
10.16 cm
29.8 cm
27 cm
26.5 cm
5.4 kg
Molded Glass
Rectangular
4 in.
11.75 in.
10.75 in.
10.47 in.
12 lb.