Learn More
Fisherbrand™ Fridge/Freezer/Room Thermometer
With freezer/room temperature display.
Merk:  Fisherbrand™ 98769-61
30.30 EUR valid until 2025-07-13
Use promo code "24135" to get your promotional price.
Produktcode. 12712277
  • 44.75 /
Only null left
Edge
Toevoegen aan winkelmandje
Beschrijving
Beschrijving
  • High/Low temperature alarm setting
  • Max./Min. temperature memory
  • Waterproof fridge sensor
  • °C/°F exchange
Specifications:
  • Measurement Range:
    • -10° to +50°C/14° to 122°F (room temperature)
    • -50° to +70°C/-58° to +158°F (fridge/freezer temperature)
  • Display Size: 26 x 12 mm
  • Product Size: 63 x 48 x 12 mm
  • Sensor cord length: 1000 mm
  • Battery: 1.5 V, size A76 or equivalent x 1 pc
  • Multi-point calibration on an individually-numbered Traceable™ Certificate which assures accuracy from an ISO/IEC 17025:2017 (1750.01) calibration laboratory accredited by A2LA
Specificatie
Specificatie
63 x 48 x 12 mm
8 cm
Plastic
Fridge/Freezer/Room Thermometer
70°C
-50°C
Room Temperature:-10°C to +50°C; Fridge to Freezer Temperature:-50°C to +70°C
Digital
26 x 12 mm
1000 mm
Waterproof
158°F
-58°F
Room Temperature: 14°F to 122°F; Fridge° to Freezer Temperature: -58°F to +158°F
No
6 cm