Learn More
DuPont™ Tychem 6000 F CHA5 Hooded Coverall
Offering excellent chemical permeation protection to an extensive range of chemicals. DuPont™ Tychem™ 6000 F Hooded Coveralls are made from a lightweight and durable fabric consisting of a proprietary barrier film-laminated to a heavy duty Tyvek substrate.
Merk: DuPont™ D13395268
35.82 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Bekijk meerdere types van dit product via de
Beschrijving
- Available in grey for discretionary needs
- Elasticated face, wrists, waist and ankles for optimal fit
- Thumb loops prevent sleeves from riding up
- Self-adhesive chin flap for tight seal of suit to face masks
- Self-adhesive double zipper flap closure system
- Stitched and over-taped seams with barrier tape for protection and strength
- Protects personnel against numerous toxic industrial organic chemicals, highly concentrated inorganic chemicals (even under pressure), particulates, radioactive contamination, biohazards and certain chemical warfare agents
- Used for a broad range of applications from chemical spill clean-up, emergency response to military and petrochemical applications
Specificatie
Hooded Coveralls | |
Tychem™-textiel | |
Elastic | |
Dubbel | |
Elastisch | |
Gestikt en afgeplakt, Taped | |
Chemicals | |
(EU) 2016/425, EN 14126, EN 1073-2, EN 1149-5 | |
Voor een breed toepassingsbereik, van het opruimen van gemorste chemische vloeistoffen en hulpverlening bij noodgevallen tot militaire en petrochemische toepassingen |
2X-Large | |
Grijs | |
Elastisch | |
Hooded | |
Rits, overslag | |
Category III, Type 3-B, 4-B, 5-B and 6-B | |
Zelfklevende kin | |
116 tot 124 cm |