Learn More
Fisherbrand™ Schilden tegen bètastraling
Ideal for protection from Beta radiation, Fisherbrand™ radiation shields are supplied in small, medium and large sizes and with a 15cm-deep curved base for use with safety trays, or a 30x30cm flat base for under-the-bench protection.
Merk:  Fisherbrand™ CSR-S2T
 Bekijk meerdere types van dit product via de
Produktcode. 12347187
  • 390.00 / 1 stuk
Only null left
Toevoegen aan winkelmandje
Beschrijving
Beschrijving
A range of angles offers increased maneuverability, while clear optical acrylic aids visualisation.
Specificatie
Specificatie
Curved Base
Large Fixed 15° Angle
Beta Radiation Shield
530 x 350 x 300 mm