Learn More
3M™ Standaard chemisch adsorptiemiddel
Absorbeer bijna elke vloeistof met deze chemische absorptiemiddelen
Merk: 3M™ 7000051994
Beschrijving
Meltblown polypropylene microfibers absorb liquids such as hydrofluoric acid and other aqueous-based chemicals, and soak up an average of 10 times their weight of most EPA-designated hazardous liquids.
- Sorbents are dirt- and dust-free; contain no abrasive granules that can damage expensive equipment
- Polypropylene base is chemically inert and incinerable; allows separation and recycling in many applications
- Easy-to-spot yellow cartons protect sorbents from temperature and humidity for secure, long-term storage
- Convenient dispenser boxes—suitable for tabletops or wall mounting—keep pads and pillows within easy reach Pads or rolls are ideal for use on work surfaces and can be trimmed to size
- Mini booms surround and contain spills; pillows soak them up
- 3M Catalog# P110, P200, P208, P300, P500
These safety products must be used in accordance with the user instructions, warnings and limitations accompanying each product.
Specificatie
OSHA 29 CFR 1910.132 | |
12 gallon | |
Polypropyleen | |
Voor beheersing van morsen van bepaalde gevaarlijke vloeistoffen | |
4 ft. | |
3 inch |
Geel | |
45.4 l | |
Minislang voor chemische absorptie | |
P200 | |
1.21 m | |
7.62 cm |