Learn More
Invitrogen™ Lipofectamine™ 2000 Transfection Reagent
Versatile transfection reagent that has been shown to effectively transfect the widest variety of adherent and suspension cell lines
Merk: Invitrogen™ 11668030
287.56 EUR valid until 2025-06-30
Use promo code "24074" to get your promotional price.
This item is not returnable.
View return policy
Beschrijving
Lipofectamine™ 2000 Transfection Reagent is a versatile transfection reagent that has been shown to effectively transfect the widest variety of adherent and suspension cell lines. Researchers use Lipofectamine™ 2000 Reagent for siRNA- and shRNA-based gene knockdown experiments, as well as for gene expression studies. Lipofectamine™ 2000 Transfection Reagent is our best transfection reagent available for the co-transfection of siRNA and plasmid DNA.With Lipofectamine™ 2000 Transfection Reagent, you'll get:
A high-performance transfection reagent for gene expression and gene silencing
Lipofectamine™ 2000 Transfection Reagent works effectively with all common cell lines as well as with many challenging ones, and can be used in media with or without serum. For gene silencing, Lipofectamine™ 2000 Transfection Reagent's high-efficiency transfections provide the high levels of gene knockdown needed to achieve convincing results. Lipofectamine™ 2000 Transfection Reagent is the number one choice for co-transfection, given its effectiveness for transfecting both siRNA and plasmid DNA. Lipofectamine™ 2000 Transfection Reagent is easy to use—simply mix with nucleic acid and add to cell culture.
Ideal for high-throughput work
Simplicity and speed combined with high transfection efficiency make Lipofectamine™ 2000 Transfection Reagent ideal for transient protein expression or high-throughput RNAi transfections. Transfection conditions can be easily established for automated or robotic systems used in such applications.
Informatie over bestelling
Sustainable packaging. Approved for shipment at Room Temperature or on Wet Ice.
Specificatie
Each unit contains one vial of Lipofectamine™ 2000 Reagent (0.3 ml). Store at 2–8°C. |
|
12-well Plate | |
Transfection | |
High-throughput Compatible | |
Transfection Reagent | |
Yes | |
Lipid-Based Transfection |
Established Cell Lines, Stem Cells, Primary Cells, Hard-to-Transfect Cells | |
Plasmid DNA, Synthetic siRNA, RNAi Plasmids (shRNA, miR) | |
Sustainable packaging | |
Lipofectamine™ | |
0.3 mL | |
Approved for shipment at Room Temperature or on Wet Ice |