Learn More
Carbolite™ CWF Chamber Furnace
Bench-mounted, general purpose laboratory chamber furnace. Carbolite™ CWF Chamber Furnace is available in a variety of sizes with a maximum operating temperature up to 1300°C.
Merk:  Carbolite™ CWF1113-230SN+&02-301
 Bekijk meerdere types van dit product via de
Produktcode. 12049842
  • 6840.00 / 1 stuk
Only null left
Toevoegen aan winkelmandje
Beschrijving
Beschrijving
  • 1100°C, 1200°C or 1300°C maximum operating temperature.
  • 5, 13, 23, 36 or 65L chamber volumes
  • Carbolite Gero 301 controller, with single ramp to set-point and process timer
  • Soft closing door on 5, 13, 21 and 23L models
  • Parallel action door keeps heated surface away from the user
  • Delayed start / process timer function as standard
  • Hard wearing alumina element carrier, furnace entrance and hearth
  • Energy efficient low thermal mass insulation
  • Free radiating wire wound elements for optimum uniformity
  • Easy access to elements and controls simplifies maintenance and servicing
  • A variety of additional options are available. These must be requested at the time of purchase.
    • Over-temperature protection (recommended to protect valuable contents and for unattended operation)
    • A range of sophisticated digital controllers, multi-segment programmers and data loggers is available. These can be fitted with RS232, RS485 or Ethernet communications
    • A range of metallic retorts to work with modified atmospheres up to 1150°C
    • AMS2750E Nadcap compatible models are available for aerospace applications.
Specificatie
Specificatie
13 L
Chamber Furnace
200 x 200 x 325 mm
47 kg
1100°C
Programmable Temperature Controller
905 x 435 x 610 mm
3100 w max., 1500 w holding