Learn More
Bufferoplossing, pH 1.679
BIOPHARMA
Reagecon's pH Buffer Solution at 25°C is tested and certified by an ISO17025 Accredited Test Method (INAB Ref: 264T) and NIST traceable, has guaranteed stability throughout its entire shelf life, even after opening the bottle.
Merk:  Reagecon 101679
 Bekijk meerdere types van dit product via de
Produktcode. 11832643
  • 28.50 /
Edge
Beschrijving
Beschrijving
To ensure reliable, reportable pH measurements, this pH buffer standard is directly traceable to the IUPAC pH scale by an unbroken chain of traceability. Traceable analysis is necessary for consistency and universal acceptance of pH results, including acceptance by regulatory bodies.
  • Calibrated to a standard precision NIST value
  • Accredited to ISO 17025
Specificatie
Specificatie
0°C
Kleurloos
Liquid
Characteristic
Fully miscible in water
100°C
1.679 ±0.01
500 ml