Learn More
3M™ Versaflo™ M-300 Series Respiratory Hardhat Assembly
Versaflo™ M-300 Series Respiratory Hardhat Assembly provides respiratory and limited head and eye protection.
Merk:  3M™ 7000104036
 Bekijk meerdere types van dit product via de
Produktcode. 11773916
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Bekijk hier alternatieve producten »
Explore more special offers
Beschrijving
Beschrijving
Features:
  • Chemical and Scratch Resistant Lenses
  • Large Viewing Area
  • Lightweight, Easy Visor Replacement System
Environmental Conditions:
  • Cold Working Environment
  • High Mobility Required
  • Safety Helmet Required
Hazard Type:
  • Gas, Vapour and Particulates
  • Liquid Splash
  • Low Energy Flying Particles/Dust
  • Falling Objects
  • Sparks and Hot Particles
Applications:
  • Automotive, food and beverage manufacturing
  • Metal production and fabrication
  • Military maintenance
  • Repair and operation
  • Pharmaceutical
  • Transportation
TRUSTED_SUSTAINABILITY
Specificatie
Specificatie
25
Black
Respirator Hard Hat
3M Air Delivery Units
Polycarbonate
Non-sterile
Premium Visor and Faceseal
346 x 346 x 254 mm
346 mm
346 mm
254 mm
832 g
CE Approved, EN 12941 TH2/TH3, EN 14594 3B, EN 166:2001, EN 397
Flame-resistant Polyester
Demolition, Food and Beverage Manufacturing, Metalworking, Pulp and Paper, Agriculture, Smelting, Woodworking, Chemical Processing, Construction
Flame-Resistant Faceseal
M-307
Textile
Yes
13.6 in.
13.6 in.
9.6 in.
1.8 lb.