Learn More
Terumo™ Driedelige injectiespuit
Use these syringes for the aspiration of fluids or sampling blood or other body fluids. Terumo™ Three-Part Syringes feature a unique double beveled design for greater patient comfort.
Merk:  Terumo™ 8SS05S21381
 Bekijk meerdere types van dit product via de
Produktcode. 11773313
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Bekijk hier alternatieve producten »
Explore more special offers
Beschrijving
Beschrijving
  • Ultra-sharp needles
  • Clear barrels
  • Bold, precise scale markings
  • Smooth plunger action
  • Tamper evident peel packs
TRUSTED_SUSTAINABILITY
Specificatie
Specificatie
Ja
21 G
38 mm
100 per pak
Ja
Polypropylene
Ja
3-delige injectiespuit
5 ml